siRNA Details
| Gene Symbol: Slc35b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106590.1 | NP_001100060.1 | 3, 4 | 522 | ||
| XM_008762768.1 | XP_008760990.1 | 2, 3 | 421 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc35b4| Gene Symbol: Slc35b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106590.1 | NP_001100060.1 | 3, 4 | 522 | ||
| XM_008762768.1 | XP_008760990.1 | 2, 3 | 421 | |||