siRNA Details
| Gene Symbol: Eif2ak3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031599.2 | NP_113787.1 | 5 | 1032 | ||
| XM_006236623.1 | XP_006236685.1 | 5 | 550 | |||
| XM_006236624.2 | XP_006236686.1 | 5 | 1070 | |||
| XM_008762977.1 | XP_008761199.1 | 4 | 664 | |||
| GenBank mRNA | AF096835.1 | - | 5 | 1033 | ||

Eif2ak3
Genomic Map