siRNA Details
Gene Symbol: Gimap6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011968.3 | NP_001011968.1 | 3 | 968 | ||
GenBank mRNA | AJ633683.1 | - | 3 | 971 | ||
BC089859.1 | - | 877 | 7384582 | |||
DQ125342.1 | - | 3 | 878 | |||
DQ125343.1 | - | 878 |
