siRNA Details
| Gene Symbol: Gabra5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017295.1 | NP_058991.1 | 6 | 613 | ||
| GenBank mRNA | X51992.1 | - | 6 | 640 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gabra5| Gene Symbol: Gabra5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017295.1 | NP_058991.1 | 6 | 613 | ||
| GenBank mRNA | X51992.1 | - | 6 | 640 | ||