siRNA Details
| Gene Symbol: Snx10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013085.1 | NP_001013103.1 | 5 | 331 | ||
| XM_006236486.2 | XP_006236548.1 | 5 | 367 | |||
| XM_006236488.2 | XP_006236550.1 | 5 | 772 | |||
| XM_006236489.2 | XP_006236551.1 | 5 | 638 | |||
| XM_006236490.1 | XP_006236552.1 | 5 | 393 | |||
| GenBank mRNA | BC091290.1 | - | 5 | 331 | 7306793 | |

Snx10
Genomic Map