siRNA Details
| Gene Symbol: Gars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC088347.1 | - | 14 | 1527 | ||
| FQ231456.1 | - | 4 | 296 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gars| Gene Symbol: Gars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC088347.1 | - | 14 | 1527 | ||
| FQ231456.1 | - | 4 | 296 | |||