siRNA Details
| Gene Symbol: Fkbp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007646.1 | NP_001007647.1 | 3 | 650 | ||
| GenBank mRNA | BC081961.1 | - | 3 | 650 | 7124723 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fkbp9| Gene Symbol: Fkbp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007646.1 | NP_001007647.1 | 3 | 650 | ||
| GenBank mRNA | BC081961.1 | - | 3 | 650 | 7124723 | |