siRNA Details
| Gene Symbol: Kcnj2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017296.1 | NP_058992.1 | 1 | 992 | ||
| XM_006247567.2 | XP_006247629.1 | 2 | 4325 | |||
| GenBank mRNA | AF021137.1 | - | 1 | 992 | ||
| L48490.1 | - | 1 | 992 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnj2| Gene Symbol: Kcnj2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017296.1 | NP_058992.1 | 1 | 992 | ||
| XM_006247567.2 | XP_006247629.1 | 2 | 4325 | |||
| GenBank mRNA | AF021137.1 | - | 1 | 992 | ||
| L48490.1 | - | 1 | 992 | |||