siRNA Details
| Gene Symbol: Hspa13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019271.2 | NP_062144.2 | 5 | 1158 | ||
| GenBank mRNA | AF006617.1 | - | 5 | 1158 | ||
| BC070954.1 | - | 5 | 1158 | 7103134 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hspa13| Gene Symbol: Hspa13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019271.2 | NP_062144.2 | 5 | 1158 | ||
| GenBank mRNA | AF006617.1 | - | 5 | 1158 | ||
| BC070954.1 | - | 5 | 1158 | 7103134 | ||