siRNA Details
| Gene Symbol: LOC103692173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763090.1 | XP_008761312.1 | 5 | 656 | ||
| XM_008763091.1 | XP_008761313.1 | 5 | 584 | |||
| XM_008763092.1 | XP_008761314.1 | 4 | 575 | |||
| XM_008763093.1 | XP_008761315.1 | 4 | 477 | |||
| Gene Symbol: NEWGENE_1562258 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763087.1 | XP_008761309.1 | 8 | 2842 | ||
| XM_008763089.1 | XP_008761311.1 | 8 | 3080 | |||
| Gene Symbol: Wbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106600.1 | NP_001100070.1 | 616 | |||
| XM_006236712.2 | XP_006236774.1 | 675 | ||||
| XM_006236713.2 | XP_006236775.1 | 584 | ||||
| XM_008763021.1 | XP_008761243.1 | 477 | ||||
| GenBank mRNA | BC079453.1 | - | 1173 | |||
| BC098709.1 | - | 4 | 1170 | |||
| BC158831.1 | - | 573 | 7379825 | |||

LOC103692173
Genomic Map