siRNA Details
| Gene Symbol: Slc4a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212512.1 | NP_997677.1 | 17 | 2241 | ||
| GenBank mRNA | AB118905.1 | - | 15 | 2005 | ||
| AY496959.1 | - | 17 | 2241 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc4a5| Gene Symbol: Slc4a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212512.1 | NP_997677.1 | 17 | 2241 | ||
| GenBank mRNA | AB118905.1 | - | 15 | 2005 | ||
| AY496959.1 | - | 17 | 2241 | |||