siRNA Details
Gene Symbol: Tex261 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017537.1 | NP_001017537.1 | 5, 6 | 500 | ||
GenBank mRNA | BC091302.1 | - | 5, 6 | 500 | 7309682 | |
FQ214220.1 | - | 5, 6 | 512 | |||
FQ218231.1 | - | 5, 6 | 511 |
