siRNA Details
| Gene Symbol: LOC103690006 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763101.1 | XP_008761323.1 | 5 | 605 | ||
| Gene Symbol: Dusp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025650.1 | NP_001020821.1 | 5 | 526 | ||
| GenBank mRNA | BC098712.1 | - | 5 | 526 | 7321787 | |

LOC103690006
Genomic Map