siRNA Details
| Gene Symbol: Podxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106607.1 | NP_001100077.1 | 3, 4 | 1137 | ||
| GenBank mRNA | BC161970.1 | - | 3, 4 | 1179 | 9028963 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Podxl2| Gene Symbol: Podxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106607.1 | NP_001100077.1 | 3, 4 | 1137 | ||
| GenBank mRNA | BC161970.1 | - | 3, 4 | 1179 | 9028963 | |