siRNA Details
| Gene Symbol: Ppp4r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106613.2 | NP_001100083.2 | 6, 7 | 710 | ||
| XM_006236962.2 | XP_006237024.1 | 6, 7 | 784 | |||
| GenBank mRNA | BC167050.1 | - | 6, 7 | 628 | 7114402 | |
| EV776660.1 | - | 3, 4 | 189 | |||

Ppp4r2
Genomic Map