siRNA Details
| Gene Symbol: Galnt7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022926.1 | NP_075215.1 | 2 | 450 | ||
| XM_006253083.1 | XP_006253145.1 | 2 | 475 | |||
| GenBank mRNA | AF076167.1 | - | 2 | 450 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Galnt7| Gene Symbol: Galnt7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022926.1 | NP_075215.1 | 2 | 450 | ||
| XM_006253083.1 | XP_006253145.1 | 2 | 475 | |||
| GenBank mRNA | AF076167.1 | - | 2 | 450 | ||