siRNA Details
| Gene Symbol: Ywhae | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031603.1 | NP_113791.1 | 5 | 719 | ||
| GenBank mRNA | BC063163.1 | - | 5 | 697 | 6921685 | |
| D30739.1 | - | 5 | 703 | |||
| FQ215270.1 | - | 3 | 377 | |||
| FQ220393.1 | - | 5 | 724 | |||
| M84416.1 | - | 5 | 699 | |||
| U53882.1 | - | 5 | 719 | |||

Ywhae
Genomic Map