siRNA Details
| Gene Symbol: St6galnac3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019123.1 | NP_061996.1 | 2, 3 | 449 | ||
| GenBank mRNA | BC072501.1 | - | 2, 3 | 286 | 7099426 | |
| L29554.1 | - | 2, 3 | 449 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

St6galnac3| Gene Symbol: St6galnac3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019123.1 | NP_061996.1 | 2, 3 | 449 | ||
| GenBank mRNA | BC072501.1 | - | 2, 3 | 286 | 7099426 | |
| L29554.1 | - | 2, 3 | 449 | |||