siRNA Details
| Gene Symbol: Pyroxd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004234.1 | NP_001004234.1 | 11, 12 | 1293 | ||
| GenBank mRNA | BC079377.1 | - | 11, 12 | 1293 | 7110714 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pyroxd1| Gene Symbol: Pyroxd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004234.1 | NP_001004234.1 | 11, 12 | 1293 | ||
| GenBank mRNA | BC079377.1 | - | 11, 12 | 1293 | 7110714 | |