siRNA Details
| Gene Symbol: Tm7sf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011970.1 | NP_001011970.1 | 11 | 1509 | ||
| GenBank mRNA | BC090030.1 | - | 11 | 1509 | 7366240 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tm7sf3| Gene Symbol: Tm7sf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011970.1 | NP_001011970.1 | 11 | 1509 | ||
| GenBank mRNA | BC090030.1 | - | 11 | 1509 | 7366240 | |