siRNA Details
| Gene Symbol: Jph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106630.1 | NP_001100100.1 | 2 | 1127 | ||
| XM_006237719.2 | XP_006237781.1 | 2 | 1127 | |||
| XM_006237720.2 | XP_006237782.1 | 2 | 1127 | |||
| XM_006237721.2 | XP_006237783.1 | 2 | 1127 | |||
| XM_006237722.2 | XP_006237784.1 | 2 | 1127 | |||
| XM_006237723.2 | XP_006237785.1 | 2 | 1127 | |||
| XM_008763475.1 | XP_008761697.1 | 2 | 1127 | |||

Jph1
Genomic Map