siRNA Details
| Gene Symbol: Ndufaf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276441.1 | NP_001263370.1 | 5, 6 | 602 | ||
| GenBank mRNA | FQ215404.1 | - | 5, 6 | 602 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ndufaf6| Gene Symbol: Ndufaf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276441.1 | NP_001263370.1 | 5, 6 | 602 | ||
| GenBank mRNA | FQ215404.1 | - | 5, 6 | 602 | ||