siRNA Details
| Gene Symbol: Tp53inp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181084.2 | NP_851598.1 | 1 | 183 | ||
| XM_006237868.2 | XP_006237930.1 | 2 | 314 | |||
| XM_008763528.1 | XP_008761750.1 | 1 | 87 | |||
| XM_008763529.1 | XP_008761751.1 | 1 | 87 | |||
| GenBank mRNA | AB107917.1 | - | 1 | 183 | ||

Tp53inp1
Genomic Map