siRNA Details
| Gene Symbol: Tp53inp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181084.2 | NP_851598.1 | 2, 3 | 594 | ||
| XM_006237868.2 | XP_006237930.1 | 3, 4 | 725 | |||
| GenBank mRNA | AB107917.1 | - | 2, 3 | 594 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tp53inp1| Gene Symbol: Tp53inp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181084.2 | NP_851598.1 | 2, 3 | 594 | ||
| XM_006237868.2 | XP_006237930.1 | 3, 4 | 725 | |||
| GenBank mRNA | AB107917.1 | - | 2, 3 | 594 | ||