siRNA Details
| Gene Symbol: Sytl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025651.1 | NP_001020822.1 | 12 | 1308 | ||
| XM_006239013.2 | XP_006239075.1 | 12 | 2146 | |||
| GenBank mRNA | BC098679.1 | - | 12 | 1308 | 7384784 | |
| FQ227771.1 | - | 3 | 284 | |||

Sytl1
Genomic Map