siRNA Details
| Gene Symbol: Otud6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106639.1 | NP_001100109.1 | 6, 7 | 999 | ||
| XM_006237914.2 | XP_006237976.1 | 6, 7 | 1029 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Otud6b| Gene Symbol: Otud6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106639.1 | NP_001100109.1 | 6, 7 | 999 | ||
| XM_006237914.2 | XP_006237976.1 | 6, 7 | 1029 | |||