siRNA Details
| Gene Symbol: Wwp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024757.1 | NP_001019928.1 | 21 | 2570 | ||
| XM_006237930.2 | XP_006237992.1 | 21 | 2485 | |||
| XM_006237931.2 | XP_006237993.1 | 20 | 2912 | |||
| XM_008763590.1 | XP_008761812.1 | 22 | 2783 | |||
| XM_008763591.1 | XP_008761813.1 | 14 | 1978 | |||
| GenBank mRNA | BC097386.1 | - | 21 | 2570 | 7446512 | |
| FQ216775.1 | - | 6 | 661 | |||

Wwp1
Genomic Map