siRNA Details
| Gene Symbol: Fhl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013088.1 | NP_001013106.1 | 6 | 797 | ||
| XM_008763594.1 | XP_008761816.1 | 6 | 1162 | |||
| XM_008763595.1 | XP_008761817.1 | 5 | 1100 | |||
| GenBank mRNA | BC079327.1 | - | 6 | 797 | 7135490 | |

Fhl5
Genomic Map