siRNA Details
| Gene Symbol: Rars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106643.2 | NP_001100113.1 | 8 | 619 | ||
| XM_006237986.2 | XP_006238048.1 | 8 | 627 | |||
| GenBank mRNA | BC158744.1 | - | 8 | 619 | 9100690 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rars2| Gene Symbol: Rars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106643.2 | NP_001100113.1 | 8 | 619 | ||
| XM_006237986.2 | XP_006238048.1 | 8 | 627 | |||
| GenBank mRNA | BC158744.1 | - | 8 | 619 | 9100690 | |