siRNA Details
| Gene Symbol: Ndufb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106646.1 | NP_001100116.1 | 2 | 325 | ||
| GenBank mRNA | FQ216998.1 | - | 2 | 252 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ndufb6| Gene Symbol: Ndufb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106646.1 | NP_001100116.1 | 2 | 325 | ||
| GenBank mRNA | FQ216998.1 | - | 2 | 252 | ||