siRNA Details
Gene Symbol: Bag1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106647.3 | NP_001100117.2 | 2, 3 | 679 | ||
NM_001256084.1 | NP_001243013.1 | 2, 3 | 679 | |||
GenBank mRNA | BC159418.1 | - | 2, 3 | 679 | 8370352 | |
BF523560.1 | - | 1, 2 | 89 | |||
BI280304.1 | - | 5, 6 | 637 | |||
DY309876.1 | - | 2, 3 | 675 | |||
FQ224976.1 | - | 2, 3 | 676 |
