siRNA Details
| Gene Symbol: Bag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106647.3 | NP_001100117.2 | 4 | 769 | ||
| NM_001256084.1 | NP_001243013.1 | 4 | 769 | |||
| GenBank mRNA | BC159418.1 | - | 4 | 769 | 8370352 | |
| BF523560.1 | - | 3 | 179 | |||
| BI280304.1 | - | 4 | 547 | |||
| FQ215132.1 | - | 4 | 731 | |||
| FQ224976.1 | - | 4 | 766 | |||

Bag1
Genomic Map