siRNA Details
| Gene Symbol: Tex10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106653.1 | NP_001100123.1 | 11 | 2548 | ||
| XM_006238034.2 | XP_006238096.1 | 10 | 3574 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tex10| Gene Symbol: Tex10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106653.1 | NP_001100123.1 | 11 | 2548 | ||
| XM_006238034.2 | XP_006238096.1 | 10 | 3574 | |||