siRNA Details
| Gene Symbol: Erp44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008317.1 | NP_001008318.1 | 3, 4 | 251 | ||
| GenBank mRNA | AY158662.1 | - | 3, 4 | 251 | ||
| FQ211910.1 | - | 345 | ||||
| FQ216126.1 | - | 3, 4 | 305 | |||
| FQ227188.1 | - | 3, 4 | 311 | |||
| FQ234126.1 | - | 3, 4 | 312 | |||
| FQ234429.1 | - | 3, 4 | 352 | |||

Erp44
Genomic Map