siRNA Details
Gene Symbol: Aldh1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011975.1 | NP_001011975.1 | 2 | 570 | ||
GenBank mRNA | BC081884.1 | - | 2 | 570 | 7108511 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Aldh1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011975.1 | NP_001011975.1 | 2 | 570 | ||
GenBank mRNA | BC081884.1 | - | 2 | 570 | 7108511 |