siRNA Details
| Gene Symbol: Zbtb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106657.1 | NP_001100127.1 | 2 | 541 | ||
| XM_006238046.2 | XP_006238108.1 | 2 | 539 | |||
| XM_006238047.2 | XP_006238109.1 | 2 | 1172 | |||
| XM_006238048.2 | XP_006238110.1 | 2 | 1822 | |||

Zbtb5
Genomic Map