siRNA Details
Gene Symbol: RGD1563975 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195270.1 | NP_001182199.1 | 1 | 876 | ||
GenBank mRNA | BC128763.1 | - | 1 | 876 | 7113360 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1563975 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195270.1 | NP_001182199.1 | 1 | 876 | ||
GenBank mRNA | BC128763.1 | - | 1 | 876 | 7113360 |