siRNA Details
Gene Symbol: Bnc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106666.1 | NP_001100136.1 | 5 | 672 | ||
XM_008763775.1 | XP_008761997.1 | 5 | 1947 | |||
XM_008763776.1 | XP_008761998.1 | 4 | 1863 | |||
XM_008763777.1 | XP_008761999.1 | 5 | 1946 |
