siRNA Details
| Gene Symbol: Stoml2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031646.1 | NP_001026816.1 | 9 | 910 | ||
| GenBank mRNA | BC099164.1 | - | 9 | 910 | 7382023 | |
| FQ213146.1 | - | 9 | 941 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stoml2| Gene Symbol: Stoml2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031646.1 | NP_001026816.1 | 9 | 910 | ||
| GenBank mRNA | BC099164.1 | - | 9 | 910 | 7382023 | |
| FQ213146.1 | - | 9 | 941 | |||