siRNA Details
| Gene Symbol: Zfp353 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_233185.7 | XP_233185.6 | 1 | 2075 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp353| Gene Symbol: Zfp353 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_233185.7 | XP_233185.6 | 1 | 2075 | ||