siRNA Details
| Gene Symbol: Pcsk9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199253.2 | NP_954862.2 | 7, 8 | 1374 | ||
| GenBank mRNA | AY847775.1 | - | 7, 8 | 1374 | ||
| BC133063.1 | - | 7, 8 | 1305 | 8371686 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcsk9| Gene Symbol: Pcsk9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199253.2 | NP_954862.2 | 7, 8 | 1374 | ||
| GenBank mRNA | AY847775.1 | - | 7, 8 | 1374 | ||
| BC133063.1 | - | 7, 8 | 1305 | 8371686 | ||