siRNA Details
| Gene Symbol: Acot11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271383.1 | NP_001258312.1 | 11, 12 | 1269 | ||
| XM_008763851.1 | XP_008762073.1 | 11, 12 | 1289 | |||
| XM_008763852.1 | XP_008762074.1 | 11, 12 | 1230 | |||

Acot11
Genomic Map