siRNA Details
| Gene Symbol: Tmem59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001139465.2 | NP_001132937.1 | 3 | 411 | ||
| GenBank mRNA | CK476149.1 | - | 3 | 416 | ||
| FM094601.1 | - | 2 | 143 | |||
| FQ212504.1 | - | 3 | 377 | |||
| FQ214069.1 | - | 3 | 408 | |||
| FQ226828.1 | - | 2 | 131 | |||
| FQ231829.1 | - | 3 | 395 | |||
| FQ232106.1 | - | 3 | 416 | |||

Tmem59
Genomic Map