siRNA Details
| Gene Symbol: Tmem59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001139465.2 | NP_001132937.1 | 5 | 649 | ||
| GenBank mRNA | CK476149.1 | - | 5 | 654 | ||
| FM091711.1 | - | 1 | 69 | |||
| FM094601.1 | - | 4 | 381 | |||
| FQ212504.1 | - | 5 | 615 | |||
| FQ214069.1 | - | 5 | 646 | |||
| FQ226828.1 | - | 4 | 369 | |||
| FQ231829.1 | - | 5 | 633 | |||
| FQ232106.1 | - | 5 | 654 | |||

Tmem59
Genomic Map