siRNA Details
| Gene Symbol: Tmem59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001139465.2 | NP_001132937.1 | 7, 8 | 870 | ||
| GenBank mRNA | BF287725.1 | - | 3, 4 | 219 | ||
| CV126271.1 | - | 1, 2 | 47 | |||
| FM091711.1 | - | 3, 4 | 290 | |||
| FM094601.1 | - | 6, 7 | 602 | |||
| FQ226828.1 | - | 6, 7 | 590 | |||
| FQ231829.1 | - | 7, 8 | 856 | |||
| FQ232106.1 | - | 7, 8 | 875 | |||

Tmem59
Genomic Map