siRNA Details
| Gene Symbol: Lrrc42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025653.1 | NP_001020824.1 | 5 | 935 | ||
| XM_006238503.2 | XP_006238565.1 | 5 | 857 | |||
| GenBank mRNA | BC098685.1 | - | 5 | 935 | 7385408 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc42| Gene Symbol: Lrrc42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025653.1 | NP_001020824.1 | 5 | 935 | ||
| XM_006238503.2 | XP_006238565.1 | 5 | 857 | |||
| GenBank mRNA | BC098685.1 | - | 5 | 935 | 7385408 | |