siRNA Details
| Gene Symbol: Ube2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013933.1 | NP_001013955.1 | 6, 7 | 505 | ||
| XM_006257446.2 | XP_006257508.1 | 5, 6 | 518 | |||
| GenBank mRNA | BC079353.1 | - | 6, 7 | 505 | 7108236 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ube2a| Gene Symbol: Ube2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013933.1 | NP_001013955.1 | 6, 7 | 505 | ||
| XM_006257446.2 | XP_006257508.1 | 5, 6 | 518 | |||
| GenBank mRNA | BC079353.1 | - | 6, 7 | 505 | 7108236 | |