siRNA Details
| Gene Symbol: Slc6a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037544.1 | NP_001032633.1 | 9 | 1328 | ||
| GenBank mRNA | AJ969954.1 | - | 9 | 1328 | ||
| EF494736.1 | - | 1300 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc6a14| Gene Symbol: Slc6a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037544.1 | NP_001032633.1 | 9 | 1328 | ||
| GenBank mRNA | AJ969954.1 | - | 9 | 1328 | ||
| EF494736.1 | - | 1300 | ||||