siRNA Details
| Gene Symbol: Osbpl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044234.2 | NP_001037699.2 | 16 | 1426 | ||
| GenBank mRNA | FQ228495.1 | - | 17 | 1482 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Osbpl9| Gene Symbol: Osbpl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044234.2 | NP_001037699.2 | 16 | 1426 | ||
| GenBank mRNA | FQ228495.1 | - | 17 | 1482 | ||