siRNA Details
| Gene Symbol: Fam221b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013934.1 | NP_001013956.1 | 2 | 451 | ||
| XM_008763669.1 | XP_008761891.1 | 2 | 661 | |||
| XM_008763670.1 | XP_008761892.1 | 2 | 661 | |||
| GenBank mRNA | BC081906.1 | - | 2 | 451 | 7111080 | |

Fam221b
Genomic Map